Assay Details
| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204298.1 | NP_989629.1 | 4 - 5 | 480 | 78 | |
| GenBank mRNA | AF004664.1 | - | 4 - 5 | 480 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIMP2| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204298.1 | NP_989629.1 | 4 - 5 | 480 | 78 | |
| GenBank mRNA | AF004664.1 | - | 4 - 5 | 480 | 78 | |
Set Membership: |
|