Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204786.1 | NP_990117.1 | 1 - 2 | 68 | 80 | |
| GenBank mRNA | D13760.1 | - | 1 - 2 | 50 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_204786.1 transcripts.
Set Membership: |
|

PPY
Genomic Map