Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204786.1 | NP_990117.1 | 2 - 3 | 247 | 84 | |
| GenBank mRNA | D13760.1 | - | 2 - 3 | 229 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPY| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204786.1 | NP_990117.1 | 2 - 3 | 247 | 84 | |
| GenBank mRNA | D13760.1 | - | 2 - 3 | 229 | 84 | |
Set Membership: |
|