Assay Details
| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204983.1 | NP_990314.1 | 2 - 3 | 458 | 89 | |
| GenBank mRNA | L40932.1 | - | 2 - 3 | 458 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFNA2| Gene Symbol: EFNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204983.1 | NP_990314.1 | 2 - 3 | 458 | 89 | |
| GenBank mRNA | L40932.1 | - | 2 - 3 | 458 | 89 | |
Set Membership: |
|