Assay Details
| Gene Symbol: MAFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205025.1 | NP_990356.1 | 1 | 1921 | 101 | |
| GenBank mRNA | AF034570.1 | - | 1 | 1921 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAFA| Gene Symbol: MAFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205025.1 | NP_990356.1 | 1 | 1921 | 101 | |
| GenBank mRNA | AF034570.1 | - | 1 | 1921 | 101 | |
Set Membership: |
|