Assay Details
| Gene Symbol: CNTFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205028.1 | NP_990359.1 | 4 - 5 | 470 | 61 | |
| XM_015277243.2 | XP_015132729.1 | 4 - 5 | 605 | 61 | ||
| XM_015277246.2 | XP_015132732.1 | 4 - 5 | 614 | 61 | ||
| GenBank mRNA | U29245.1 | - | 2 - 3 | 311 | 61 | |
| Z48168.1 | - | 470 | 61 | |||
More Information
Set Membership: |
|

CNTFR
Genomic Map