Assay Details
| Gene Symbol: AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205109.2 | NP_990440.1 | 3 - 4 | 105 | 104 | |
| XM_015279336.1 | XP_015134822.1 | 3 - 4 | 104 | |||
| XM_025156030.1 | XP_025011798.1 | 3 - 4 | 104 | |||
| XM_025156031.1 | XP_025011799.1 | 3 - 4 | 104 | |||
| GenBank mRNA | BX933779.1 | - | 3 - 4 | 104 | ||
| BX935116.1 | - | 3 - 4 | 104 | |||
| CR407140.1 | - | 3 - 4 | 104 | |||
| M12153.1 | - | 3 - 4 | 108 | 104 | ||
More Information
Set Membership: |
|

AK1
Genomic Map