Assay Details
| Gene Symbol: CCNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205239.2 | NP_990570.2 | 8 - 9 | 775 | 70 | |
| XM_015278199.2 | XP_015133685.1 | 8 - 9 | 818 | 70 | ||
| XM_015278200.2 | XP_015133686.1 | 8 - 9 | 825 | 70 | ||
| GenBank mRNA | X75757.1 | - | 722 | 70 | ||
More Information
Set Membership: |
|

CCNB3
Genomic Map