Assay Details
| Gene Symbol: NFIX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205270.1 | NP_990601.1 | 9 - 10 | 1413 | 65 | |
| XM_025145699.1 | XP_025001467.1 | 10 - 11 | 1597 | 65 | ||
| XM_025145700.1 | XP_025001468.1 | 9 - 10 | 1471 | 65 | ||
| XM_025145701.1 | XP_025001469.1 | 9 - 10 | 1365 | 65 | ||
| XM_025145702.1 | XP_025001470.1 | 8 - 9 | 1305 | 65 | ||
| GenBank mRNA | X61225.1 | - | 1413 | 65 | ||
More Information
Set Membership: |
|

NFIX
Genomic Map