Assay Details
| Gene Symbol: MT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205275.1 | NP_990606.1 | 1 - 2 | 82 | 99 | |
| GenBank mRNA | X06749.1 | - | 82 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205275.1, X06749.1 transcripts.
Set Membership: |
|

MT4
Genomic Map