Assay Details
| Gene Symbol: BZFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205309.3 | NP_990640.3 | 1 - 2 | 111 | 98 | |
| GenBank mRNA | X15538.1 | - | 91 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205309.3, X15538.1 transcripts.
Set Membership: |
|

BZFP1
Genomic Map