Assay Details
| Gene Symbol: THRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205313.2 | NP_990644.1 | 7 - 8 | 858 | 58 | |
| GenBank mRNA | Y00987.1 | - | 859 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THRA| Gene Symbol: THRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205313.2 | NP_990644.1 | 7 - 8 | 858 | 58 | |
| GenBank mRNA | Y00987.1 | - | 859 | 58 | ||
Set Membership: |
|