Assay Details
| Gene Symbol: BGLAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205387.3 | NP_990718.2 | 1 - 2 | 81 | 109 | |
| GenBank mRNA | U10578.1 | - | 1 - 2 | 81 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205387.3 transcripts.
Set Membership: |
|

BGLAP
Genomic Map