Assay Details
| Gene Symbol: CNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205516.1 | NP_990847.1 | 5 - 6 | 505 | 59 | |
| XM_015302686.2 | XP_015158172.1 | 5 - 6 | 528 | 59 | ||
| GenBank mRNA | M63559.1 | - | 5 - 6 | 505 | 59 | |
More Information
Set Membership: |
|

CNN1
Genomic Map