Assay Details
| Gene Symbol: BVES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001299.2 | NP_001001299.2 | 7 - 8 | 1332 | 62 | |
| GenBank mRNA | AF124511.1 | - | 7 - 8 | 1333 | 62 | |
| AF208398.1 | - | 7 - 8 | 1302 | 62 | ||
More Information
Set Membership: |
|

BVES
Genomic Map