Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012693.1 | NP_001012711.1 | 1 - 2 | 205 | 112 | |
| XM_025148586.1 | XP_025004354.1 | 1 - 2 | 3311 | 114 | ||
| GenBank mRNA | AJ851531.1 | - | 1 - 2 | 205 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ851531.1, NM_001012693.1, XM_025148586.1 transcripts.
Set Membership: |
|

MGAT3
Genomic Map