Assay Details
| Gene Symbol: PRLHR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024585.2 | NP_001019756.2 | 1 | 1021 | 108 | |
| XM_015286221.2 | XP_015141707.1 | 2 | 7364 | 108 | ||
| XM_015286222.2 | XP_015141708.1 | 2 | 7259 | 108 | ||
| GenBank mRNA | EU117423.1 | - | 1 | 1021 | 108 | |
More Information
Set Membership: |
|

PRLHR2
Genomic Map