Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177738.1 | NP_001171209.1 | 4 - 5 | 1274 | 76 | |
| GenBank mRNA | GU907099.1 | - | 4 - 5 | 1274 | 76 | |
More Information
Set Membership: |
|

FSCN2
Genomic Map