Assay Details
| Gene Symbol: PLA2G2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184949.1 | NP_001171878.1 | 1 - 2 | 35 | 100 | |
| XM_025142544.1 | XP_024998312.1 | 2 - 3 | 2396 | 100 | ||
| GenBank mRNA | GU474517.1 | - | 1 - 2 | 35 | 100 | |
More Information
Set Membership: |
|

PLA2G2E
Genomic Map