Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194983.1 | NP_001181912.1 | 1 - 2 | 121 | 67 | |
| XM_015279464.1 | XP_015134950.1 | 2 - 3 | 300 | 67 | ||
| XM_015279465.2 | XP_015134951.1 | 2 - 3 | 199 | 67 | ||
| XM_015279466.2 | XP_015134952.1 | 2 - 3 | 285 | 67 | ||
| XM_015279467.2 | XP_015134953.1 | 2 - 3 | 209 | 67 | ||
| XM_015279468.2 | XP_015134954.1 | 2 - 3 | 313 | 67 | ||
| XM_015279469.2 | XP_015134955.1 | 2 - 3 | 287 | 67 | ||
| XM_015279470.2 | XP_015134956.1 | 2 - 3 | 234 | 67 | ||
| XM_015279472.2 | XP_015134958.1 | 2 - 3 | 409 | 67 | ||
| XM_025156072.1 | XP_025011840.1 | 2 - 3 | 234 | 67 | ||
| XM_025156073.1 | XP_025011841.1 | 2 - 3 | 253 | 67 | ||
| GenBank mRNA | BX934520.1 | - | 1 - 2 | 152 | 67 | |
| HM584710.1 | - | 1 - 2 | 121 | 67 | ||
More Information
Set Membership: |
|

PTGES
Genomic Map