Assay Details
| Gene Symbol: SDR42E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195157.1 | NP_001182086.1 | 10 - 11 | 1204 | 144 | |
| XM_015294226.2 | XP_015149712.1 | 13 - 14 | 1586 | 144 | ||
| XM_015294227.2 | XP_015149713.1 | 11 - 12 | 2015 | 144 | ||
| XM_025155108.1 | XP_025010876.1 | 13 - 14 | 1861 | 144 | ||
| XM_025155109.1 | XP_025010877.1 | 10 - 11 | 2003 | 144 | ||
More Information
Set Membership: |
|

SDR42E2
Genomic Map