Assay Details
| Gene Symbol: SDR42E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195157.1 | NP_001182086.1 | 8 - 9 | 1077 | 105 | |
| XM_015294226.2 | XP_015149712.1 | 11 - 12 | 1459 | 105 | ||
| XM_015294227.2 | XP_015149713.1 | 9 - 10 | 1888 | 105 | ||
| XM_025155108.1 | XP_025010876.1 | 11 - 12 | 1734 | 105 | ||
| XM_025155109.1 | XP_025010877.1 | 8 - 9 | 1876 | 105 | ||
More Information
Set Membership: |
|

SDR42E2
Genomic Map