Assay Details
| Gene Symbol: ELOVL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197310.1 | NP_001184239.1 | 1 - 2 | 233 | 78 | |
| XM_015277347.2 | XP_015132833.1 | 2 - 3 | 147 | 78 | ||
| XM_015277348.2 | XP_015132834.1 | 2 - 3 | 227 | 78 | ||
| XM_025144756.1 | XP_025000524.1 | 2 - 3 | 261 | 78 | ||
| GenBank mRNA | HQ264104.1 | - | 1 - 2 | 233 | 78 | |
More Information
Set Membership: |
|

ELOVL7
Genomic Map