Assay Details
| Gene Symbol: LYRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198888.1 | NP_001185817.1 | 2 - 3 | 341 | 131 | |
| XM_015275893.2 | XP_015131379.1 | 2 - 3 | 255 | 131 | ||
More Information
Set Membership: |
|

LYRM4
Genomic Map