Assay Details
| Gene Symbol: DPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199164.1 | NP_001186093.1 | 3 - 4 | 214 | 94 | |
| GenBank mRNA | AJ720186.1 | - | 3 - 4 | 206 | 94 | |
| CR390371.1 | - | 1 - 2 | 65 | 94 | ||
More Information
Set Membership: |
|

DPM2
Genomic Map