Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199391.1 | NP_001186320.1 | 1 - 2 | 99 | 104 | |
| GenBank mRNA | CB018491.1 | - | 1 - 2 | 99 | 104 | |
More Information
Set Membership: |
|

MAL
Genomic Map