Assay Details
| Gene Symbol: ERGIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199403.1 | NP_001186332.1 | 9 - 10 | 692 | 84 | |
| XM_015275706.2 | XP_015131192.1 | 9 - 10 | 789 | 84 | ||
| XM_015275707.2 | XP_015131193.1 | 9 - 10 | 825 | 84 | ||
| XM_015275708.2 | XP_015131194.1 | 9 - 10 | 716 | 84 | ||
| GenBank mRNA | BU369329.1 | - | 1 - 2 | 58 | 84 | |
| CR352981.1 | - | 2 - 3 | 144 | 84 | ||
More Information
Set Membership: |
|

ERGIC2
Genomic Map