Assay Details
| Gene Symbol: METTL15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199422.1 | NP_001186351.1 | 1 - 2 | 288 | 77 | |
| XM_015286163.2 | XP_015141649.1 | 2 - 3 | 857 | 77 | ||
| XM_025150639.1 | XP_025006407.1 | 2 - 3 | 857 | 77 | ||
| XM_025150640.1 | XP_025006408.1 | 2 - 3 | 298 | 77 | ||
| XM_025150641.1 | XP_025006409.1 | 2 - 3 | 490 | 77 | ||
| XM_025150642.1 | XP_025006410.1 | 2 - 3 | 857 | 77 | ||
| XM_025150643.1 | XP_025006411.1 | 2 - 3 | 857 | 77 | ||
| XM_025150644.1 | XP_025006412.1 | 2 - 3 | 298 | 77 | ||
| GenBank mRNA | BM491151.1 | - | 1 - 2 | 288 | 77 | |
More Information
Set Membership: |
|

METTL15P1
Genomic Map