Assay Details
| Gene Symbol: METTL15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199422.1 | NP_001186351.1 | 3 - 4 | 607 | 66 | |
| XM_015286163.2 | XP_015141649.1 | 4 - 5 | 1176 | 66 | ||
| XM_025150639.1 | XP_025006407.1 | 5 - 6 | 1200 | 66 | ||
| XM_025150640.1 | XP_025006408.1 | 5 - 6 | 641 | 66 | ||
| XM_025150641.1 | XP_025006409.1 | 5 - 6 | 833 | 66 | ||
| XM_025150642.1 | XP_025006410.1 | 5 - 6 | 1200 | 66 | ||
| XM_025150644.1 | XP_025006412.1 | 4 - 5 | 617 | 66 | ||
| GenBank mRNA | BM491151.1 | - | 3 - 4 | 607 | 66 | |
More Information
Set Membership: |
|

METTL15P1
Genomic Map