Assay Details
| Gene Symbol: METTL15P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199422.1 | NP_001186351.1 | 4 - 5 | 796 | 68 | |
| XM_015286163.2 | XP_015141649.1 | 5 - 6 | 1365 | 68 | ||
| XM_025150639.1 | XP_025006407.1 | 6 - 7 | 1389 | 68 | ||
| XM_025150640.1 | XP_025006408.1 | 6 - 7 | 830 | 68 | ||
| XM_025150641.1 | XP_025006409.1 | 6 - 7 | 1022 | 68 | ||
More Information
Set Membership: |
|

METTL15P1
Genomic Map