Assay Details
| Gene Symbol: ZC3HC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199437.1 | NP_001186366.1 | 9 - 10 | 1381 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC3HC1| Gene Symbol: ZC3HC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199437.1 | NP_001186366.1 | 9 - 10 | 1381 | 62 | |
Set Membership: |
|