Assay Details
| Gene Symbol: PMPCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199472.1 | NP_001186401.1 | 2 - 3 | 245 | 63 | |
| XM_015275727.2 | XP_015131213.1 | 2 - 3 | 257 | 63 | ||
| GenBank mRNA | AM064989.1 | - | 2 - 3 | 245 | 63 | |
More Information
Set Membership: |
|

PMPCB
Genomic Map