Assay Details
| Gene Symbol: GNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199559.1 | NP_001186488.1 | 5 - 6 | 692 | 71 | |
| GenBank mRNA | CF256802.1 | - | 1 - 2 | 73 | 71 | |
More Information
Set Membership: |
|

GNS
Genomic Map