Assay Details
| Gene Symbol: SPAG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199586.1 | NP_001186515.1 | 4 - 5 | 466 | 60 | |
| XM_015281383.2 | XP_015136869.1 | 4 - 5 | 600 | 60 | ||
| XM_025147182.1 | XP_025002950.1 | 3 - 4 | 2437 | 60 | ||
| XM_025147183.1 | XP_025002951.1 | 5 - 6 | 612 | 60 | ||
More Information
Set Membership: |
|

SPAG6
Genomic Map