Assay Details
| Gene Symbol: SLC16A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199604.1 | NP_001186533.1 | 3 - 4 | 1173 | 98 | |
| XM_015281443.2 | XP_015136929.1 | 4 - 5 | 1436 | 98 | ||
| XM_015281459.2 | XP_015136945.1 | 4 - 5 | 1248 | 98 | ||
| XM_025147871.1 | XP_025003639.1 | 3 - 4 | 5411 | 98 | ||
More Information
Set Membership: |
|

SLC16A7
Genomic Map