Assay Details
| Gene Symbol: TOMM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199617.1 | NP_001186546.1 | 2 - 3 | 555 | 60 | |
| GenBank mRNA | BU301105.1 | - | 2 - 3 | 384 | 60 | |
| BU364982.1 | - | 2 - 3 | 554 | 60 | ||
| CN232573.1 | - | 2 - 3 | 142 | 60 | ||
More Information
Set Membership: |
|

TOMM7
Genomic Map