Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199643.3 | NP_001186572.2 | 4 - 5 | 1166 | 61 | |
| XM_015286998.2 | XP_015142484.1 | 4 - 5 | 1194 | 61 | ||
More Information
Set Membership: |
|

SYT13
Genomic Map