Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199701.1 | NP_001186630.1 | 1 - 2 | 204 | 60 | |
| XM_015287338.2 | XP_015142824.1 | 1 - 2 | 241 | 60 | ||
| GenBank mRNA | BM440475.1 | - | 1 - 2 | 204 | 60 | |
| CN217066.1 | - | 1 - 2 | 174 | 60 | ||
More Information
Set Membership: |
|

PIGH
Genomic Map