Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199701.1 | NP_001186630.1 | 2 - 3 | 409 | 71 | |
| GenBank mRNA | BM440475.1 | - | 2 - 3 | 409 | 71 | |
| CN217066.1 | - | 2 - 3 | 379 | 71 | ||
More Information
Set Membership: |
|

PIGH
Genomic Map