Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199702.1 | NP_001186631.1 | 2 - 3 | 764 | 63 | |
| XM_015281499.2 | XP_015136985.1 | 4 - 5 | 1080 | 63 | ||
| XM_015281500.2 | XP_015136986.1 | 4 - 5 | 1207 | 63 | ||
More Information
Set Membership: |
|

SNRK
Genomic Map