Assay Details
| Gene Symbol: CDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199857.1 | NP_001186786.1 | 6 - 7 | 794 | 52 | |
| GenBank mRNA | EF182713.1 | - | 6 - 7 | 794 | 52 | |
More Information
Set Membership: |
|

CDK2
Genomic Map