Assay Details
| Gene Symbol: CD38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201388.1 | NP_001188317.1 | 3 - 4 | 565 | 86 | |
| GenBank mRNA | HM769338.1 | - | 3 - 4 | 565 | 86 | |
More Information
Set Membership: |
|

CD38
Genomic Map