Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204815.1 | NP_001191744.1 | 3 - 4 | 302 | 114 | |
| XM_015290917.2 | XP_015146403.1 | 3 - 4 | 364 | 114 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map