Assay Details
| Gene Symbol: TMEM189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205100.1 | NP_001192029.1 | 2 - 3 | 395 | 78 | |
| GenBank mRNA | CR387613.1 | - | 2 - 3 | 395 | 78 | |
More Information
Set Membership: |
|

TMEM189
Genomic Map