Assay Details
| Gene Symbol: RAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243724.1 | NP_001230653.1 | 1 - 2 | 442 | 101 | |
| GenBank mRNA | AB020318.1 | - | 1 - 2 | 275 | 101 | |
| AF420600.1 | - | 1 - 2 | 442 | 101 | ||
More Information
Set Membership: |
|

RAX
Genomic Map