Assay Details
| Gene Symbol: GPR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256138.1 | NP_001243067.1 | 1 - 2 | 398 | 69 | |
| GenBank mRNA | JF795914.1 | - | 1 - 2 | 398 | 69 | |
| JF795915.1 | - | 1 - 2 | 398 | 69 | ||
More Information
Set Membership: |
|

GPR83
Genomic Map