Assay Details
| Gene Symbol: TIA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257203.1 | NP_001244132.1 | 10 - 11 | 1023 | 70 | |
| XM_015297483.2 | XP_015152969.2 | 10 - 11 | 1234 | 70 | ||
| XM_025142788.1 | XP_024998556.1 | 11 - 12 | 1195 | 70 | ||
| XM_025142789.1 | XP_024998557.1 | 9 - 10 | 836 | 70 | ||
| XM_025142790.1 | XP_024998558.1 | 7 - 8 | 632 | 70 | ||
| XM_025142791.1 | XP_024998559.1 | 5 - 6 | 505 | 70 | ||
| XM_025142792.1 | XP_024998560.1 | 11 - 12 | 1098 | 70 | ||
| GenBank mRNA | AY189903.1 | - | 10 - 11 | 859 | 70 | |
| BU434325.1 | - | 3 - 4 | 295 | 70 | ||
More Information
Set Membership: |
|

TIA1
Genomic Map