Assay Details
| Gene Symbol: SERPINF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257289.1 | NP_001244218.1 | 2 - 3 | 151 | 61 | |
| XM_015295746.2 | XP_015151232.1 | 3 - 4 | 1698 | 61 | ||
| XM_025141957.1 | XP_024997725.1 | 3 - 4 | 1694 | 61 | ||
| GenBank mRNA | BU130808.1 | - | 2 - 3 | 130 | 61 | |
| BX935009.2 | - | 2 - 3 | 134 | 61 | ||
| BX950646.1 | - | 2 - 3 | 151 | 61 | ||
| CR387489.1 | - | 2 - 3 | 150 | 61 | ||
More Information
Set Membership: |
|

SERPINF1
Genomic Map