Assay Details
| Gene Symbol: FBXL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270994.1 | NP_001257923.1 | 2 - 3 | 308 | 70 | |
| XM_025154853.1 | XP_025010621.1 | 2 - 3 | 298 | 70 | ||
| GenBank mRNA | AJ451550.1 | - | 2 - 3 | 295 | 70 | |
| BU306928.1 | - | 2 - 3 | 253 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270994.1, XM_025154853.1 transcripts.
Set Membership: |
|

FBXL21
Genomic Map