Assay Details
| Gene Symbol: FBXL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270994.1 | NP_001257923.1 | 4 - 5 | 541 | 184 | |
| NM_001270995.1 | NP_001257924.1 | 3 - 4 | 364 | 184 | ||
| GenBank mRNA | AJ451550.1 | - | 4 - 5 | 528 | 184 | |
| BU306928.1 | - | 4 - 5 | 486 | 184 | ||
| DR427211.1 | - | 3 - 4 | 325 | 184 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270994.1, NM_001270995.1 transcripts.
Set Membership: |
|

FBXL21
Genomic Map